Merck
  • Spectrofluorometric measurements of the dispersion state of pyrenedodecanoic acid and its uptake by cultured cells and liposomes.
Spectrofluorometric measurements of the dispersion state of pyrenedodecanoic acid and its uptake by cultured cells and liposomes.
Biochimica et biophysica acta (1988-09-01)
S Gatt, E Fibach
ABSTRACT
Pyrene dodecanoic acid (P12), a medium-chain fatty acid to which the fluorescent probe pyrene is covalently linked, showed a considerable increase in fluorescence when the probe was introduced into a hydrophobic environment. Also, when closely packed in an aggregate, an energy transfer between two adjacent molecules of pyrene occurred, resulting in a shift of the peak of the emission spectrum from 378 nm ('monomeric') to 475 nm ('excimeric'). These two respective properties were utilized for the following: (a) A spectrofluorometric measurement of the critical micellar concentration (CMC) of the pyrene fatty acid, defined as the concentration at which the 475 nm emission peak appeared as a consequence of the aggregation of P12 molecules in aqueous solution to form micelles; the CMC of P12 was found to be in the range of 1 to 2 microM. (b) The penetration of P12, from an aqueous solution or dispersion, into unilamellar phospholipid vesicles was determined by monitoring the increase of the fluorescence at 378 nm. The fluorescence increase was time-dependent and proportional to the respective concentrations of P12 or phospholipid vesicles. Substituting the neutral phosphatidylcholine with the negatively-charged phosphatidylserine vesicles resulted in a slower rate as well as lesser total uptake of P12. (c) The uptake of P12 by cells was accompanied by an increase in the monomeric fluorescence emission intensity. Using cells in suspension, this could be followed continuously in a spectrofluorometer equipped with a recorder. The uptake was found to be time-dependent and proportional to P12 concentration.