Merck
  • Combined inhibition of serotonin uptake and oxidative deamination attenuates audiogenic seizures in DBA/2J mice.
Combined inhibition of serotonin uptake and oxidative deamination attenuates audiogenic seizures in DBA/2J mice.
Pharmacology, biochemistry, and behavior (1985-11-01)
D L Sparks, N S Buckholtz
ABSTRACT
We previously reported that 6-methoxy-1,2,3,4-tetrahydro-beta-carboline (6-MeO-THBC) attenuated audiogenic seizures (AGS) in 21-day-old DBA/2J mice and also inhibited brain monoamine oxidase-A (MAO-A) and serotonin (5-hydroxytryptamine, 5-HT) uptake leading to increased brain 5-HT concentration. In this study, the sensitivity of AGS to 5-HT manipulation was evaluated by utilizing drug combinations which paralleled the actions of 6-MeO-THBC and which also have been associated with the production of a serotonergic motor syndrome in rats. Combination of a specific 5-HT uptake inhibitor (fluoxetine or citalopram) with the MAO-A inhibitor clorgyline inhibited AGS more effectively than the individual drugs but combination with the MAO-B inhibitor deprenyl did not. Combined administration of clorgyline plus deprenyl also suppressed AGS. Inhibition of AGS by tryptophan was potentiated by combination with either of the mixed MAO inhibitors nialamide or tranylcypromine. The effects of these drugs individually and in combination on brain MAO-A and MAO-B activity and 5-HT uptake were also determined ex vivo and were consistent with expected mechanisms of action. These results suggest, first of all, that the inhibition of AGS produced by 6-MeO-THBC is a consequence of its combined MAO-A and 5-HT uptake inhibition properties. Secondly, the similarity of results of pharmacological manipulations of the 5-HT system which produce the rat motor syndrome and which inhibit AGS in the mouse suggests that AGS in 21-day-old DBA/2J mice may be a useful system for assessing functional consequences of these serotonergic manipulations.