Merck
The clinical use of arginine aspartate in male infertility.
Acta Europaea fertilitatis (1982-09-01)
D De Aloysio, R Mantuano, M Mauloni, G Nicoletti
ABSTRACT
130 male subjects (61 with asthenospermia, 69 with oligoasthenospermia) classified according to spermiologic patterns have been treated with different therapeutic outlines (gonadotrophins, antibiotica, arginine aspartate high levels, their associations); 89 subjects have been undergone with arginine aspartate, 41 without arginine aspartate. The results have been critically examined in order to different therapeutic outlines, to spermiogram, to diagnosis, to use of arginine aspartate in different spermiologic alterations, to use of arginine aspartate with respect to control groups, to arginine aspartate levels.