Merck
  • [Determination of furadan and its metabolites in biomedia by thin-layer chromatography].
[Determination of furadan and its metabolites in biomedia by thin-layer chromatography].
Gigiena i sanitariia (1982-07-01)
V N Os'kina