Merck
  • Residues of the plant growth regulator mefluidide [N-[2,4]dimethy-5-[[(trifluoromethyl)sulfonyl]amino]phenyl]acetamide] in the milk and tissues of lactating dairy cows: a 28-day feeding study.
Residues of the plant growth regulator mefluidide [N-[2,4]dimethy-5-[[(trifluoromethyl)sulfonyl]amino]phenyl]acetamide] in the milk and tissues of lactating dairy cows: a 28-day feeding study.
Journal of agricultural and food chemistry (1981-11-01)
D E Clark, C E Coppock, G W Ivie
Product Description
Supelco
Mefluidide, PESTANAL®, analytical standard