Merck
  • Role of the ubiquitin-proteasome pathway in regulating abundance of the cyclin-dependent kinase inhibitor p27.
Role of the ubiquitin-proteasome pathway in regulating abundance of the cyclin-dependent kinase inhibitor p27.
Science (New York, N.Y.) (1995-08-04)
M Pagano, S W Tam, A M Theodoras, P Beer-Romero, G Del Sal, V Chau, P R Yew, G F Draetta, M Rolfe
ABSTRACT
The p27 mammalian cell cycle protein is an inhibitor of cyclin-dependent kinases. Both in vivo and in vitro, p27 was found to be degraded by the ubiquitin-proteasome pathway. The human ubiquitin-conjugating enzymes Ubc2 and Ubc3 were specifically involved in the ubiquitination of p27. Compared with proliferating cells, quiescent cells exhibited a smaller amount of p27 ubiquitinating activity, which accounted for the marked increase of p27 half-life measured in these cells. Thus, the abundance of p27 in cells is regulated by degradation. The specific proteolysis of p27 may represent a mechanism for regulating the activity of cyclin-dependent kinases.