Merck
  • Inhibitory effect of ONO-1078 on specific binding of peptide leukotrienes to human lung crude membrane.
Inhibitory effect of ONO-1078 on specific binding of peptide leukotrienes to human lung crude membrane.
Prostaglandins, leukotrienes, and essential fatty acids (1994-09-01)
T Nabe, S Kohno, T Tanpo, Y Saeki, H Yamamura, M Horiba, K Ohata
ABSTRACT
We investigated the effects of ONO-1078, a newly synthesized peptide leukotriene (p-LT antagonist, on the specific binding of radiolabelled [3H]-LTC4, [3H]-LTD4 and [3H]-LTE4 to a human lung crude membrane fraction (HLMF). The binding assay was performed under conditions in which [3H]-LTC4 and [3H]-LTD4 were not metabolized by HLMF; that is, the metabolism of LTC4 to LTD4 or LTE4 was almost completely prevented by pretreating HLMF with 5 mM acivicin at 37 degrees C for 180 min, and metabolism of LTD4 to LTE4 was inhibited by including 5 mM L-cysteine and 5 mM glycine in the assay. [3H]-LTD4 specific binding was potently and concentration-dependently dissociated by ONO-1078. Its potency was 180-fold stronger than that of FPL 55712, a standardized p-LT antagonist, whereas high concentrations of ONO-1078 similar to those of FPL 55712 were required to inhibit [3H]-LTC4 specific binding. The rank order of the inhibitory potencies of p-LT agonists and antagonists for [3H]-LTD4 specific binding was LTD4 > ONO-1078 > LTE4 > LTC4 > FPI 55712. On the other hand, not only high concentrations of ONO-1078 and FPL 55712 but also more than a 100-fold excess of unlabelled LTE4 was required to inhibit [3H]-LTE4 specific binding, indicating that the binding sites do not appear to be receptors of LTE4. From these results, it is suggested that ONO-1078 is a highly potent LTD4 antagonist which is expected to be very effective on bronchial asthma.