Merck
  • Biological monitoring of exposure to organophosphorus insecticides by assay of urinary alkylphosphates: influence of protective measures during manual operations with treated plants.
Biological monitoring of exposure to organophosphorus insecticides by assay of urinary alkylphosphates: influence of protective measures during manual operations with treated plants.
International archives of occupational and environmental health (1994-01-01)
C Aprea, G Sciarra, P Sartorelli, E Desideri, R Amati, E Sartorelli
ABSTRACT
Biological monitoring was carried out by assaying urinary dimethylated alkylphosphates [dimethyldithiophosphate (DMDTP), dimethylthio-phosphate (DMTP), and dimethylphosphate (DMP)] in 11 workers exposed to chlorpyrifos-methyl and azinphosmethyl during operations in a previously sprayed peach orchard. The subjects were divided into groups on the basis of the protective clothing worn. The results were compared with those of a reference group of 99 subjects not occupationally exposed to organophosphorus insecticides. The hand-wash liquid of the workers was also analyzed to evaluate skin contamination. Significantly higher levels of urinary excretion of alkylphosphates were found in all groups than in unexposed controls (Student's t test). A good correlation was found between quantities of the active ingredients on the hands and urinary excretion of total dimethylated alkylphosphates (r = 0.788) and of DMTP (r = 0.749) and DMP (r = 0.790) alone. The correlation between azinphos-methyl on the hands and urinary excretion of DMDTP was poor (r = 0.069). Under the working conditions investigated, the main route of absorption seems to be via the skin. Respiratory absorption, however, also appears significant in view of the difference in urinary excretion of dimethylated alkylphosphates found between subjects with and without face masks.