Merck
Chemoenzymatic access to enantiomeric bicyclo[2.2.1]heptan-2,5-diones.
Bioorganic & medicinal chemistry (1994-06-01)
A Weissfloch, R Azerad
ABSTRACT
A practical integrated process, combining an enzymatic resolution step with a few chemical transformations, is described for the synthesis of (1R, 4R)- and (2S, 4S)-bicyclo[2.2.1]heptan-2,5-diones 1 of high enantiomeric purity, starting from a standard mixture of (+/-)-endo- and exo-2-acetoxy-5-norbornene.