Merck
  • Dental anxiety and the absorption of orally administered erythromycin stearate.
Dental anxiety and the absorption of orally administered erythromycin stearate.
Oral surgery, oral medicine, oral pathology, oral radiology, and endodontics (1995-12-01)
W A Coulter, J G McGimpsey, A Coffey, J Strawbridge, G J Linden
ABSTRACT
Erythromycin stearate is an acid labile antibiotic, therefore fear and apprehension, which are known to affect gastric motility, may produce erratic absorption resulting in lower serum levels. The mean (SD) serum erythromycin concentration 75 minutes after a 1.5 gm oral dose of erythromycin stearate to 45 patients was 8.7 (4.8) mg/L and ranged widely from 0.4 to 20.5 mg/L. The serum concentration of erythromycin was below therapeutic levels (1.0 mg/L) in two patients. No significant association was found between anxiety and serum levels of erythromycin when age, gender, and gastric distress were taken into account. It is concluded that dental anxiety may indirectly influence the uptake of oral erythromycin stearate; but this relationship is complex, and there is no evidence from this study that increased dental anxiety decreases the uptake of the drug.