Merck
  • Cytosolic and mitochondrial enzymes inactivation by uroporphyrin in light and darkness.
Cytosolic and mitochondrial enzymes inactivation by uroporphyrin in light and darkness.
Journal of enzyme inhibition (1996-01-01)
S G Afonso, R Enriquez de Salamanca, A M Batlle
ABSTRACT
The effect of uroporphyrin I (UI) on several cytosolic and mitochondrial enzymes (succinyl CoA synthetase, delta-aminolevulinic acid synthetase, rhodanese, lactate dehydrogenase) has been examined. All the enzymes were inactivated in the presence of the porphyrin both in the dark and under UV light.