Merck
  • The synthesis of 4-arylsulfanyl-substituted kainoid analogues from trans-4-hydroxy-L-proline.
The synthesis of 4-arylsulfanyl-substituted kainoid analogues from trans-4-hydroxy-L-proline.
Bioorganic & medicinal chemistry letters (2000-09-15)
J E Baldwin, G J Pritchard, D S Williamson
ABSTRACT
The potent neuroexcitatory activity of kainoid amino acids in the mammalian CNS places new analogues in high demand as tools for neuropharmacological research. A range of 4-arylsulfanyl-substituted kainoids has been synthesised in a parallel fashion via mesylate displacement by a number of aromatic and heteroaromatic thiolates upon (2S,3S,4R)-1-benzoyl-3-tert-butoxycarbonylmethyl-4-methanesulfo nyloxy pyrrolidine-2-carboxylic acid methyl ester 8, which is obtainable in eight steps from trans-4-hydroxy-L-proline 5.
Sigma-Aldrich
trans-4-Hydroxy-L-proline, BioReagent, suitable for cell culture, ≥98.5%
Sigma-Aldrich
trans-4-Hydroxy-L-proline, ≥99%
Sigma-Aldrich
trans-4-Hydroxy-L-proline, BioXtra, ≥99.0% (NT)
Supelco
trans-4-Hydroxy-L-proline, analytical standard