Merck
  • A micro-autoanalytic procedure developed for the determination of ketone bodies, gluconeogenic amino acids, pyruvate, lactate, and glucose in metabolic studies.
A micro-autoanalytic procedure developed for the determination of ketone bodies, gluconeogenic amino acids, pyruvate, lactate, and glucose in metabolic studies.
Biochemical medicine (1975-10-11)
P T Ozand, R L Hawkins, R M Collins, J T Tildon, M Cornblath
Sigma-Aldrich
(R)-3-Hydroxybutyric acid, ≥98.0% (T)