Merck
Anti-tumor promoting effect of glycosides from Prunus persica seeds.
Biological & pharmaceutical bulletin (2003-02-11)
Toshiyuki Fukuda, Hideyuki Ito, Teruo Mukainaka, Harukuni Tokuda, Hoyoku Nishino, Takashi Yoshida
ABSTRACT
Four minor components, along with the major cyanogenic glycosides, amygdalin and prunasin, were isolated from Prunus persica seeds (Persicae Semen; Tounin), and characterized as mandelic acid glycosides (beta-gentiobioside and beta-D-glucoside) and benzyl alcohol glycosides (beta-gentiobioside and beta-D-glucoside). The anti-tumor promoting activity of these compounds was examined in both in vitro and in vivo assays. All of the compounds significantly inhibited the Epstein-Barr virus early antigen activation induced by tumor promoter. In addition, they produced a delay of two-stage carcinogenesis on mouse skin that was comparable in potency to (-)-epigallocatechin gallate from green tea. Structure-activity relationships indicated that a substituent at the benzylic position with glycosidic linkage affected the in vitro and in vivo activities with an order of enhancing potency, CN<COOH<H.
Sigma-Aldrich
Mandelic acid, 99%
Supelco
Benzyl alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Benzyl alcohol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Benzyl alcohol, anhydrous, 99.8%
Sigma-Aldrich
Benzyl alcohol, ReagentPlus®, ≥99%
Supelco
Benzyl alcohol, analytical standard
Sigma-Aldrich
Benzyl alcohol, puriss. p.a., ACS reagent, ≥99.0% (GC)
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%
Sigma-Aldrich
Benzyl alcohol, natural, ≥98%, FG
Sigma-Aldrich
Benzyl alcohol, ≥99%, FCC, FG
Supelco
DL-Mandelic acid, analytical standard