Merck
  • Identification of two novel nonsense mutations in the transglutaminase 1 gene in a Hungarian patient with congenital ichthyosiform erythroderma.
Identification of two novel nonsense mutations in the transglutaminase 1 gene in a Hungarian patient with congenital ichthyosiform erythroderma.
Experimental dermatology (2003-06-26)
K Becker, M Csikós, M Sárdy, Z S Szalai, A Horváth, S Kárpáti
ABSTRACT
Congenital ichthyosiform erythroderma (CIE) belongs together with lamellar ichthyosis (LI) to the group of autosomal recessive congenital ichthyoses (ARCI). Mutations in the transglutaminase (TGase) 1 gene (TGM1) have been identified in several families with LI and in some families with CIE. We report a case of CIE with two new nonsense mutations: a C7780G transversion in exon 11 resulting in a premature stop codon at aminoacid residue Y503X and a C8533G transversion in exon 13 leading to a nonsense mutation at S669X. These mutations were also identified in a heterozygous pattern in the unaffected parents. These two termination-codons result in the translation of a truncated protein at the C-terminal end domain of the TGM 1 molecule. B.C1 monoclonal antibody failed to detect TGase 1 in the patient's skin sample, and TGase activity measured by monodansyl cadaverine-incorporation showed the reduced TGase activity at the distribution of TGase 1 in the epidermis.
Sigma-Aldrich
Cadaverine, 95%
Sigma-Aldrich
Cadaverine, purum, ≥97.0% (GC)