Merck
  • Fatal sodium valproate-induced hypersensitivity syndrome with lichenoid dermatitis and fulminant hepatitis.
Fatal sodium valproate-induced hypersensitivity syndrome with lichenoid dermatitis and fulminant hepatitis.
Journal of the American Academy of Dermatology (2003-08-02)
Yau-Li Huang, Hong-Shang Hong, Zen-Whe Wang, Tseng-tong Kuo
ABSTRACT
Anticonvulsant hypersensitivity syndrome caused by sodium valproate (Depakene) has rarely been reported. Here we describe a patient who presented with nonspecific polymorphous eruptions, fulminant hepatitis, and jaundice. Lichenoid dermatitis was found in the skin biopsy specimen. Toxic hepatitis and microvesicular steatosis were found in the liver biopsy specimen, consistent with valproate hepatotoxicity.
Product Description
Supelco
Valproic acid solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Valproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propylpentanoic acid