Merck
  • Methionine sulfoxide and proteolytic cleavage contribute to the inactivation of cathepsin G by hypochlorous acid: an oxidative mechanism for regulation of serine proteinases by myeloperoxidase.
Methionine sulfoxide and proteolytic cleavage contribute to the inactivation of cathepsin G by hypochlorous acid: an oxidative mechanism for regulation of serine proteinases by myeloperoxidase.
The Journal of biological chemistry (2005-06-22)
Baohai Shao, Abderrazzaq Belaaouaj, Christophe L M J Verlinde, Xiaoyun Fu, Jay W Heinecke
ABSTRACT
Using myeloperoxidase and hydrogen peroxide, activated neutrophils produce high local concentrations of hypochlorous acid (HOCl). They also secrete cathepsin G, a serine protease implicated in cytokine release, receptor activation, and degradation of tissue proteins. Isolated cathepsin G was inactivated by HOCl but not by hydrogen peroxide in vitro. We found that activated neutrophils lost cathepsin G activity by a pathway requiring myeloperoxidase, suggesting that oxidants generated by myeloperoxidase might regulate cathepsin G activity in vivo. Tandem mass spectrometric analysis of oxidized cathepsin G revealed that loss of a peptide containing Asp108, which lies in the active site, associated quantitatively with loss of enzymatic activity. Catalytic domain peptides containing Asp108 were lost from the oxidized protein in concert with the conversion of Met110 to the sulfoxide. Release of this peptide was blocked by pretreating cathepsin G with phenylmethylsulfonyl fluoride, strongly implying that oxidation introduced proteolytic cleavage sites into cathepsin G. Model system studies demonstrated that methionine oxidation can direct the regiospecific proteolysis of peptides by cathepsin G. Thus, oxidation of Met110 may contribute to cathepsin G inactivation by at least two distinct mechanisms. One involves direct oxidation of the thioether residue adjacent to the aspartic acid in the catalytic domain. The other involves the generation of new sites that are susceptible to proteolysis by cathepsin G. These observations raise the possibility that oxidants derived from neutrophils restrain pericellular proteolysis by inactivating cathepsin G. They also suggest that methionine oxidation could render cathepsin G susceptible to autolytic cleavage. Myeloperoxidase may thus play a previously unsuspected role in regulating tissue injury by serine proteases during inflammation.
Product Description
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
SAFC
L-Aspartic acid
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)