Merck
  • Plasma free amino acids and their metabolites in Taiwanese patients on hemodialysis and continuous ambulatory peritoneal dialysis.
Plasma free amino acids and their metabolites in Taiwanese patients on hemodialysis and continuous ambulatory peritoneal dialysis.
Clinica chimica acta; international journal of clinical chemistry (2005-08-10)
Chih-Kuang Chuang, Shuan-Pei Lin, Han-Hsiang Chen, Yi-Chou Chen, Tuan-Jen Wang, Wen-Hsiang Shieh, Chih-Jen Wu
ABSTRACT
The high prevalence of protein-energy malnutrition is a critical issue for patients with end stage renal disease (ESRD) on hemodialysis (HD) or continuous ambulatory peritoneal dialysis (CAPD). Levels of plasma and intracellular amino acids are significant indicators of protein metabolism and nutritional status assessment. We measured plasma FAAs in patients on maintenance dialysis and to provide information in monitoring the therapeutic strategy, particularly in AA supplementary therapy or protein restriction. Fifty-five patients with ESRD were investigated, 25 on HD (male : female=14 : 11; 48-67 y) and 30 on CAPD (male : female=17 : 13; 45-64 y). The subjects had been on dialysis for an average of 13 months (range, 9 to 22 months). Their plasma FAAs (including their intermediate metabolites) were measured by ion exchange chromatography before and after HD or during CAPD and were compared with data obtained from 20 age- and sex-matched healthy controls. The total plasma FAA levels (urea and free ammonia, NH3 were excluded) in pre-HD samples (3911 +/- 709 micromol/l) was significantly higher than in the other groups (2570 +/- 378 in control, 3210 +/- 640 in post-HD, and 3468 +/- 271 in CAPD samples). The mean plasma FAA concentrations differed significantly between pre-HD and controls and between pre-HD and CAPD samples (p<0.05). No significant differences were found among the other group comparisons. Comparing individual FAA concentrations, only citrulline differed significantly among all groups (p<0.05), whereas serine, glutamine, beta-alanine, beta-aminoisobutyric acid, and gamma-aminobutyric acid were not different. Concentrations of some FAAs involved in the urea cycle, e.g., arginine, aspartic acid, citrulline, and ornithines, and solutes urea and NH3, were significantly increased. Ratios of tyrosine/phenylalanine and valine/glycine ratios were markedly reduced in all patients on dialysis compared with controls. FAAs either from dietary uptake or protein catabolism are substantially retained in the plasma of patients with ESRD, possibly producing higher levels of the waste products (urea and NH3) through the urea cycle and ammonia metabolism in liver. Maintenance dialysis can effectively eliminate excess FAAs in plasma, as there was a 17.9% reduction post-HD. The abnormalities in FAA metabolism found in patients with ESRD necessitate careful consideration of dialysis and dietary measures.
Sigma-Aldrich
Cadaverine, 95%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn2+ in THF, highly reactive Rieke®metal
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Trimethylamine solution, 43.0-49.0% in H2O (T)
Sigma-Aldrich
Trimethylamine solution, 31-35 wt. % in ethanol, 4.2 M, contains toluene as stabilizer
Sigma-Aldrich
Cadaverine, purum, ≥97.0% (GC)
Supelco
Acetone, analytical standard
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Trimethylamine solution, 25 wt. % in H2O
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Trimethylamine N-oxide, 95%
Sigma-Aldrich
4-Heptanone, 98%
Sigma-Aldrich
Zinc, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
1-Methyl-L-histidine, ≥98.0% (TLC)