Merck
  • Unchanged serum levels of advanced glycation endproducts in patients with liver disease.
Unchanged serum levels of advanced glycation endproducts in patients with liver disease.
Naunyn-Schmiedeberg's archives of pharmacology (2007-06-16)
Moritz Butscheid, Christian Schäfer, Stefanie Brenner, Dominik Alscher, Thomas Mürdter, Toshimitsu Niwa, Matthias Frischmann, Monika Pischetsrieder, Ulrich Klotz
ABSTRACT
Advanced glycation end products (AGEs), e.g., carboxymethyllysine (CML) or imidazolone are involved in several age-related disorders. Concerning their accumulation, the importance of hepatic and renal function is controversially discussed. To test whether impairment of hepatic or renal function will affect their accumulation, both AGEs have been measured in various populations, such as 52 patients with liver disease [viral hepatitis C without (n = 19) and with (n = 10) fatty liver; nonalcoholic fatty liver (n = 13), nonalcoholic steatohepatitis (n = 10)]. Serum concentrations of both AGEs have been compared to those in 20 healthy controls and 24 patients with moderate renal impairment (creatinine clearance 23-55 ml/min). Concerning CML (95% C.I. 803-1200 ng/ml), no differences between the various groups could be observed. Likewise, serum levels of imidazolone (95% C.I. 1.3-5.6 units) were similar in all populations. In conclusion, moderate impairment in hepatic or in renal function did not affect serum levels of CML and imidazolone. Apparently, any increase observed in severe cirrhosis or renal failure seems to be rather a consequence than a cause of both disorders.
Sigma-Aldrich
Zinc, nanopowder, 40-60 nm avg. part. size, ≥99% trace metals basis
Sigma-Aldrich
Zinc, shot, 5 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, foil, thickness 1.0 mm, 99.99% trace metals basis
Sigma-Aldrich
Trimethylamine solution, 25 wt. % in H2O
Sigma-Aldrich
Zinc, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Zinc, sticks, diam. 7-10 mm, 99.97% trace metals basis
Sigma-Aldrich
Zinc, pieces, 2-14 mesh, 99.9% trace metals basis
Sigma-Aldrich
Zinc, dust, <10 μm, ≥98%
Sigma-Aldrich
Zinc, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.9%, granular
Sigma-Aldrich
Trimethylamine solution, 31-35 wt. % in ethanol, 4.2 M, contains toluene as stabilizer
Sigma-Aldrich
Zinc preparation, 5 g/dL Zn2+ in THF, highly reactive Rieke®metal
Sigma-Aldrich
Trimethylamine solution, 43.0-49.0% in H2O (T)
Sigma-Aldrich
Zinc, shot, <12 mm, 99.99% trace metals basis
Sigma-Aldrich
Zinc, granular, 20-30 mesh, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Cadaverine, 95%
Sigma-Aldrich
Cadaverine, purum, ≥97.0% (GC)
Sigma-Aldrich
Dimethylamine solution, 2.0 M in THF
Sigma-Aldrich
Dimethylamine solution, 2.0 M in methanol
Sigma-Aldrich
Zinc, mossy, ≥99%
Sigma-Aldrich
Dimethylamine solution, purum, 33% in absolute ethanol (~5.6 M)
Sigma-Aldrich
Inosine, ≥99% (HPLC)