Merck
Urinary mandelic acid as an exposure test for ethylbenzene.
International archives of occupational and environmental health (1984-01-01)
J P Gromiec, J K Piotrowski
ABSTRACT
Absorption of ethylbenzene and excretion of mandelic acid were investigated under controlled conditions in six volunteers, exposed at concentrations of 18, 34, 80, and 200 mg/m3. Retention of ethylbenzene vapours in the lungs was 49 +/- 5%. Elimination of mandelic acid was found to be biphasic, with biological half-life values of 3.1 and 24.5 h. Total excreted mandelic acid accounts for 55 +/- 2% of retained ethylbenzene. The results obtained were applied to devise an exposure test for ethylbenzene, which would enable the precise evaluation of exposure at low ethylbenzene, vapour concentrations (+/- 13%). Exposures, carried out dermally, gave a rationale for the exclusion of the skin as a route of entry of ethylbenzene vapours into the body.
Product Description
Supelco
DL-Mandelic acid
Sigma-Aldrich
Mandelic acid, 99%