Merck
  • Sequential one-pot ruthenium-catalyzed azide-alkyne cycloaddition from primary alkyl halides and sodium azide.
Sequential one-pot ruthenium-catalyzed azide-alkyne cycloaddition from primary alkyl halides and sodium azide.
The Journal of organic chemistry (2011-03-11)
Johan R Johansson, Per Lincoln, Bengt Nordén, Nina Kann
ABSTRACT
An experimentally simple sequential one-pot RuAAC reaction, affording 1,5-disubstituted 1H-1,2,3-triazoles in good to excellent yields starting from an alkyl halide, sodium azide, and an alkyne, is reported. The organic azide is formed in situ by treating the primary alkyl halide with sodium azide in DMA under microwave heating. Subsequent addition of [RuClCp*(PPh(3))(2)] and the alkyne yielded the desired cycloaddition product after further microwave irradiation.
Sigma-Aldrich
3-Methoxybenzyl bromide, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
1-Ethynyl-4-fluorobenzene, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
3-Ethynylpyridine, 98%
Pricing and availability is not currently available.