Merck
IX. Note on Silver Foil in Surgery.
Annals of surgery (1909-10-01)
J S Lewis
Product Description
Silver, foil, 150x150mm, thickness 0.15mm, as rolled, 99.95+%
Silver, foil, 150x150mm, thickness 0.1mm, annealed, 99.95+%
Silver, foil, 50x50mm, thickness 0.1mm, annealed, 99.95+%
Silver, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99+%
Silver, foil, 25x25mm, thickness 0.125mm, as rolled, 99.95+%
Silver, foil, 10mm disks, thickness 0.025mm, as rolled, 99.99+%
Silver, foil, 150x150mm, thickness 0.20mm, as rolled, 99.95+%
Silver, foil, 0.2m coil, thickness 0.01mm, 99.95+%
Silver, foil, 25x25mm, thickness 1.0mm, as rolled, 99.95+%
Silver, foil, 10mm disks, thickness 0.02mm, 99.97%
Silver, foil, 0.5m coil, thickness 0.035mm, as rolled, 99.95+%
Silver, foil, 10mm disks, thickness 0.20mm, as rolled, 99.95+%
Silver, foil, 0.5m coil, thickness 0.025mm, annealed, 99.95+%
Silver, foil, 50x50mm, thickness 0.25mm, as rolled, 99.95+%
Silver, foil, 25x25mm, thickness 2.0mm, as rolled, 99.95+%
Silver, foil, 50x50mm, thickness 0.20mm, as rolled, 99.95+%
Silver, foil, light tested, 50x50mm, thickness 0.0125mm, 99.95+%
Silver, foil, not light tested, 300x300mm, thickness 0.025mm, as rolled, 99.95+%
Silver, foil, not light tested, 100x100mm, thickness 0.0025mm, 99.97%
Silver, foil, not light tested, 100x100mm, thickness 0.0125mm, 99.95+%
Silver, foil, 4mm disks, thickness 0.01mm, 99.95+%
Silver, foil, 25mm disks, thickness 1.0mm, as rolled, 99.99+%
Silver, foil, 6mm disks, thickness 0.005mm, 99.97%
Silver, foil, 4mm disks, thickness 0.004mm, 99.97%
Silver, foil, 25mm disks, thickness 1.0mm, as rolled, 99.95+%
Silver, foil, 2m coil, thickness 0.0125mm, 99.95+%
Silver, foil, 25mm disks, thickness 0.1mm, annealed, 99.95+%
Silver, foil, 6mm disks, thickness 0.02mm, 99.97%
Silver, foil, 50mm disks, thickness 0.008mm, 99.95+%
Silver, foil, light tested, 100x100mm, thickness 0.02mm, 99.97%