Merck
  • Cytotoxicity of N-dodecanoyl-Lhomoserine lactone and 5-N-dodecyl resorcinol to human granulocytes and monocytes: a comparative study.
Cytotoxicity of N-dodecanoyl-Lhomoserine lactone and 5-N-dodecyl resorcinol to human granulocytes and monocytes: a comparative study.
Central European Journal of Immunology (2013)
Sviridova, T. et al.