Merck
  • Metabolism of l-Threonic Acid in Rumex x acutus L. and Pelargonium crispum (L.) L'Hér.
Metabolism of l-Threonic Acid in Rumex x acutus L. and Pelargonium crispum (L.) L'Hér.
Plant physiology (1982-06-01)
J P Helsper, F A Loewus
ABSTRACT
l-Threonic acid is a natural constituent in leaves of Pelargonium crispum (L.) L'Hér (lemon geranium) and Rumex x acutus L. (sorrel). In both species, l-[(14)C]threonate is formed after feeding l-[U-(14)C]ascorbic acid to detached leaves. R. acutus leaves labeled with l-[4-(3)H]- or l-[6-(3)H]ascorbic acid produce l-[(3)H]threonate, in the first case internally labeled and in the second case confined to the hydroxymethyl group. These results are consistent with the formation of l-threonate from carbons three through six of l-ascorbic acid. Detached leaves of P. crispum oxidize l-[U-(14)C] threonate to l-[(14)C]tartrate whereas leaves of R. acutus produce negligible tartrate and the bulk of the (14)C appears in (14)CO(2), [(14)C]sucrose, and other products of carbohydrate metabolism. R. acutus leaves that are labeled with l-[U-(14)C]threonate release (14)CO(2) at linear rate until a limiting value of 25% of the total [U-(14)C]threonate is metabolized. A small quantity of [(14)C]glycerate is also produced which suggests a process involving decarboxylation of l-[U-(14)C]threonate.