Skip to Content
PL
EN
Products
Applications
Services
Resources
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Lab Automation
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Documents
Learning Center
New
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Home
Search Results
Arsenic: an old enemy now turned friend.
Arsenic: an old enemy now turned friend.
Leukemia & lymphoma (2013-04-05)
Frank Firkin, Harry Iland
PMID
23550992
Product Description
311383
Sigma-Aldrich
Arsenic(III) oxide, ACS reagent (primary standard)
Pricing
17971
Supelco
Arsenic(III) oxide, reference material for titrimetry, certified by BAM, >99.5%
Pricing
202673
Sigma-Aldrich
Arsenic(III) oxide, 99.995% trace metals basis
Pricing
A1010
Sigma-Aldrich
Arsenic(III) oxide,
ReagentPlus
®
, ≥99.0%
Pricing