Merck
  • Combined effect of glycation and sodium carbonate-bicarbonate buffer concentration on IgG binding, IgE binding and conformation of ovalbumin.
Combined effect of glycation and sodium carbonate-bicarbonate buffer concentration on IgG binding, IgE binding and conformation of ovalbumin.
Journal of the science of food and agriculture (2013-04-05)
Xiao-juan Ma, Jin-yan Gao, Hong-bing Chen
ABSTRACT
Ovalbumin (OVA) is a major allergen in hen egg. During thermal processing, reducing sugars contained in the hen egg white might easily undergo glycation with OVA, but few studies have been conducted on its corresponding immunoreactivity changes. The aim of the present study was to assess changes of the antigenicity, potential allergenicity and conformation of OVA after glycation in a wet-thermal processing system under different concentrations of sodium carbonate-bicarbonate buffer. IgE binding of the glycated OVA was increased after glycation, and the higher the sodium carbonate-bicarbonate buffer concentration, the higher the IgE binding capacity. The increase in IgE binding of OVA corresponded well with the disruption of the disulfide bond, which exposed the epitopes initially buried. Antigenicity of the glycated OVA was increased, and the amount of the increase varied among samples treated under different buffer concentrations. Glycation increased the allergenic potential for OVA, with the amount of increase varying with different sodium carbonate-bicarbonate buffer concentrations.
Product Description
Supelco
Sodium bicarbonate concentrate, 0.1 M NaHCO3 in water, eluent concentrate for IC
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate solution, solution (7.5%), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium carbonate, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Supelco
Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Supelco
Sodium carbonate concentrate, Na2CO3 72 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 7