Merck
  • Changes in the phospholipid fatty acid composition of the lipid droplet during the differentiation of 3T3-L1 adipocytes.
Changes in the phospholipid fatty acid composition of the lipid droplet during the differentiation of 3T3-L1 adipocytes.
Journal of biochemistry (2013-06-14)
Kotoko Arisawa, Ikuyo Ichi, Yukiko Yasukawa, Yasuko Sone, Yoko Fujiwara
ABSTRACT
Lipid droplets (LDs) are independent organelles in adipocytes that are composed of a lipid ester core surrounded by a phospholipid monolayer. The fatty acid composition of the phospholipid monolayer should determine the metabolism and dynamics of LDs. In this study, we examined the fatty acid composition of phospholipid monolayer in LDs during the differentiation of 3T3-L1 adipocytes. The levels of saturated fatty acids (SFAs), such as 16:0 and 18:0, in phosphatidylcholine (PC) and phosphatidylethanolamine (PE) of LDs decreased during differentiation. In contrast, the levels of monounsaturated fatty acids (MUFAs) such as 16:1n-7 and 18:1n-9 in PC and PE of LDs and the level of polyunsaturated fatty acids (PUFAs) such as 20:3n-6, 20:4n-6 and 22:6n-3 in PE of LDs increased during differentiation. These results suggest that the phospholipid monolayer in mature LDs is more fluid than that in nascent LDs. The fatty acid compositions of the LD monolayer were different from those of the microsome bilayer in the early stage of differentiation, but similar to those of the microsome bilayer in the late stage of differentiation. These data provide evidence that biophysical properties of the phospholipid monolayer in LDs change during adipocyte differentiation.
Sigma-Aldrich
L-α-Phosphatidylethanolamine from egg yolk, Type III, 10 mg/mL in chloroform, ≥97%, solution
Sigma-Aldrich
L-α-Phosphatidylethanolamine from Glycine max (soybean), Type IV, ≥98% (TLC), lyophilized powder
Sigma-Aldrich
L-α-Phosphatidylethanolamine from egg yolk, Type III, ≥97% (TLC), lyophilized powder