Merck
  • Randomized trial of clozapine vs. risperidone in treatment-naïve first-episode schizophrenia: results after one year.
Randomized trial of clozapine vs. risperidone in treatment-naïve first-episode schizophrenia: results after one year.
Schizophrenia research (2013-07-23)
Javier Sanz-Fuentenebro, Diana Taboada, Tomás Palomo, María Aragües, Santiago Ovejero, Cristina Del Alamo, Vicente Molina
ABSTRACT
In first-episode patients with psychosis, clozapine may be potentially valuable as an initial treatment seeking to limit early on clinical and cognitive deterioration. Nevertheless, until recently its restricted use has limited the study of this possibility. Our research group is developing a non-commercial, multicentric and open label study on the differential efficacy between clozapine and risperidone in first-episode schizophrenia. In this paper, we present the results related to clinical variables after a one-year follow-up. So far, we have recruited 30 patients diagnosed with schizophrenia or schizophreniform disorder with illness duration of less than two years. The patients had not received any previous treatment and they were randomized to treatment with clozapine or risperidone. Our results indicate that on average, patients on clozapine adhered to their original treatment for a longer time period than patients on risperidone. By last observation carried forward (LOCF) analysis, patients on clozapine and risperidone displayed similar clinical improvements, although marginally greater improvements in positive and total symptoms scores were found in the clozapine group. At the 12-month point we observed a marginal improvement in negative symptom scores in patients on clozapine. Subjective secondary effects, as measured with the Udvalg for KliniskeUndersøgelser (UKU) scale, correlated negatively with negative symptoms at follow-up. Our data, although preliminary, suggest that clozapine may have a slightly superior efficacy in the initial year of treatment of first-episode treatment-naïve patients with schizophrenia, and this can be explained for the most part by greater adherence to this treatment.
Product Description
Risperidone, European Pharmacopoeia (EP) Reference Standard
Clozapine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Clozapine
Sigma-Aldrich
Risperidone, ≥98% (HPLC), powder
Supelco
Risperidone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Clozapine for peak identification, European Pharmacopoeia (EP) Reference Standard
Risperidone for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Clozapine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®