Merck
  • Sorption of carbamazepine from water by magnetic molecularly imprinted polymers based on chitosan-Fe₃O₄.
Sorption of carbamazepine from water by magnetic molecularly imprinted polymers based on chitosan-Fe₃O₄.
Carbohydrate polymers (2013-08-06)
Ya-Lei Zhang, Juan Zhang, Chao-Meng Dai, Xue-Fei Zhou, Shu-Guang Liu
ABSTRACT
A novel magnetic-molecularly imprinted polymer (MMIP) based on chitosan-Fe₃O₄ has been synthesized for fast separation of carbamazepine (CBZ) from water. During polymerization, the modified chitosan-Fe₃O₄ was used not only as supporter but also as functional monomer. The properties of obtained MMIP were characterized by scanning electron and transmission electron microscopy, X-ray diffraction, Fourier transform infrared spectra, thermo-gravimetric analysis and so on. The sorption equilibrium data was well described by Freundlich isotherm model and the increase in the temperature generated an increase in the sorption amount, indicating endothermic nature of adsorption process. Sorption kinetics followed the pseudo-second-order model. The feasibility of selective sorption of CBZ from real water by the MMIP was analyzed by using spiked real water samples. The result showed that the sorption capacity of MMIP has no obvious decrease in different water samples whereas there was obvious decline in the sorption amount of the MNIP.
Sigma-Aldrich
Chitosan from shrimp shells, low-viscous
Sigma-Aldrich
Iron(III) oxide, ≥99.995% trace metals basis
Sigma-Aldrich
Iron(III) oxide, nanopowder, <50 nm particle size
Supelco
Carbamazepine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Iron(III) oxide, powder, <5 μm, ≥96%
Sigma-Aldrich
Iron(III) oxide, dispersion, nanoparticles, ≤110 nm particle size, 15 wt. % in ethanol
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chitosan, from shrimp shells, ≥75% (deacetylated)
Sigma-Aldrich
Chitosan, low molecular weight
Sigma-Aldrich
Chitosan, medium molecular weight
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Chitosan, high molecular weight
Sigma-Aldrich
Carbamazepine, powder
Sigma-Aldrich
Chitosan, from shrimp shells, practical grade