Merck
  • Septo-hippocampal GABAergic signaling across multiple modalities in awake mice.
Septo-hippocampal GABAergic signaling across multiple modalities in awake mice.
Nature neuroscience (2013-08-06)
Patrick Kaifosh, Matthew Lovett-Barron, Gergely F Turi, Thomas R Reardon, Attila Losonczy
ABSTRACT
Hippocampal interneurons receive GABAergic input from the medial septum. Using two-photon Ca(2+) imaging of axonal boutons in hippocampal CA1 of behaving mice, we found that populations of septo-hippocampal GABAergic boutons were activated during locomotion and salient sensory events; sensory responses scaled with stimulus intensity and were abolished by anesthesia. We found similar activity patterns among boutons with common putative postsynaptic targets, with low-dimensional bouton population dynamics being driven primarily by presynaptic spiking.
Sigma-Aldrich
PSEM89S Hydrochloride salt, ≥98% (HPLC)