Merck
  • Concise synthesis of the multiply oxygenated ABC-ring system of the dihydro-β-agarofurans.
Concise synthesis of the multiply oxygenated ABC-ring system of the dihydro-β-agarofurans.
Organic letters (2013-08-14)
Tomochika Ishiyama, Daisuke Urabe, Hiroki Fujisawa, Masayuki Inoue
ABSTRACT
The multiply oxygenated ABC-ring system of the dihydro-β-agarofurans was synthesized by employing two highly stereoselective reactions. The quinidine-catalyzed Diels-Alder reaction between a chiral dienophile and 3-hydroxy-4-methyl-2-pyrone simultaneously installed the C2-stereogenic center and two contiguous tetrasubstituted carbon centers (C5 and C10) of the A-ring. After 12 additional transformations, the aldol reaction of the resulting spiral AC-ring cyclized the B-ring with stereoselective introduction of the C7- and C8-centers.
Sigma-Aldrich
Maltol, natural, FG
Sigma-Aldrich
3-Hydroxy-2-methyl-4-pyrone, 99%
Sigma-Aldrich
Maltol, ≥99.0%, FCC, FG
Sigma-Aldrich
Quinidine, anhydrous
Sigma-Aldrich
Quinidine, crystallized, ≥98.0% (dried material, NT)