Merck
  • Luteibaculum oceani gen. nov., sp. nov., a carotenoid-producing, lipolytic bacterium isolated from surface seawater, and emended description of the genus Owenweeksia Lau et al. 2005.
Luteibaculum oceani gen. nov., sp. nov., a carotenoid-producing, lipolytic bacterium isolated from surface seawater, and emended description of the genus Owenweeksia Lau et al. 2005.
International journal of systematic and evolutionary microbiology (2013-08-31)
Mariyam Shahina, Asif Hameed, Shih-Yao Lin, Wei-An Lai, You-Cheng Liu, Yi-Han Hsu, Chiu-Chung Young
ABSTRACT
A yellow-pigmented, Gram-staining-negative, strictly aerobic, rod-shaped, non-flagellated, non-spore-forming, lipolytic and gliding marine bacterium designated strain CC-AMWY-103B(T) was isolated from surface seawater collected at Kending, Taiwan. The strain shared the highest 16S rRNA gene sequence similarity of 89.4% with Owenweeksia hongkongensis JCM 12287(T) and Brumimicrobium mesophilum YH207(T), and established a distinct phyletic lineage associated with the members of the family Cryomorphaceae. The polar lipid profile of strain CC-AMWY-103B(T) consisted of phosphatidylethanolamine, ten unidentified lipids and four unidentified aminolipids. The major fatty acids (>5% of the total) were iso-C15 : 0, iso-C15 : 1 G, C15 : 1ω5c, iso-C17 : 0 3-OH and C15 : 1ω8c. The DNA G+C content was 44.2 ± 0.3 mol%. The predominant respiratory quinone was menaquinone-6 (MK-6) and the major polyamine was spermidine. Based on its genetic, phylogenetic, phenotypic and chemotaxonomic distinctiveness, strain CC-AMWY-103B(T) is proposed to represent a distinct member of the family Cryomorphaceae, for which the name Luteibaculum oceani gen. nov., sp. nov. is proposed; the type strain of Luteibaculum oceani is CC-AMWY-103B(T) ( = JCM 18817(T) = BCRC 80551(T)). An emended description of the genus Owenweeksia is also proposed.
Sigma-Aldrich
L-α-Phosphatidylethanolamine from egg yolk, Type III, ≥97% (TLC), lyophilized powder
Sigma-Aldrich
Spermidine, ≥99% (GC)
Sigma-Aldrich
L-α-Phosphatidylethanolamine from egg yolk, Type III, 10 mg/mL in chloroform, ≥97%, solution
Sigma-Aldrich
L-α-Phosphatidylethanolamine from Glycine max (soybean), Type IV, ≥98% (TLC), lyophilized powder
Sigma-Aldrich
Spermidine 0.1 M solution
Sigma-Aldrich
Spermidine, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Spermidine, BioReagent, Molecular Biology, suitable for cell culture, ≥98%
Supelco
Spermidine, analytical standard