Merck
  • Rabbit 3-hydroxyhexobarbital dehydrogenase is a NADPH-preferring reductase with broad substrate specificity for ketosteroids, prostaglandin D₂, and other endogenous and xenobiotic carbonyl compounds.
Rabbit 3-hydroxyhexobarbital dehydrogenase is a NADPH-preferring reductase with broad substrate specificity for ketosteroids, prostaglandin D₂, and other endogenous and xenobiotic carbonyl compounds.
Biochemical pharmacology (2013-09-03)
Satoshi Endo, Toshiyuki Matsunaga, Atsuko Matsumoto, Yuki Arai, Satoshi Ohno, Ossama El-Kabbani, Kazuo Tajima, Yasuo Bunai, Shigeru Yamano, Akira Hara, Yukio Kitade
ABSTRACT
3-Hydroxyhexobarbital dehydrogenase (3HBD) catalyzes NAD(P)⁺-linked oxidation of 3-hydroxyhexobarbital into 3-oxohexobarbital. The enzyme has been thought to act as a dehydrogenase for xenobiotic alcohols and some hydroxysteroids, but its physiological function remains unknown. We have purified rabbit 3HBD, isolated its cDNA, and examined its specificity for coenzymes and substrates, reaction directionality and tissue distribution. 3HBD is a member (AKR1C29) of the aldo-keto reductase (AKR) superfamily, and exhibited high preference for NADP(H) over NAD(H) at a physiological pH of 7.4. In the NADPH-linked reduction, 3HBD showed broad substrate specificity for a variety of quinones, ketones and aldehydes, including 3-, 17- and 20-ketosteroids and prostaglandin D₂, which were converted to 3α-, 17β- and 20α-hydroxysteroids and 9α,11β-prostaglandin F₂, respectively. Especially, α-diketones (such as isatin and diacetyl) and lipid peroxidation-derived aldehydes (such as 4-oxo- and 4-hydroxy-2-nonenals) were excellent substrates showing low K(m) values (0.1-5.9 μM). In 3HBD-overexpressed cells, 3-oxohexobarbital and 5β-androstan-3α-ol-17-one were metabolized into 3-hydroxyhexobarbital and 5β-androstane-3α,17β-diol, respectively, but the reverse reactions did not proceed. The overexpression of the enzyme in the cells decreased the cytotoxicity of 4-oxo-2-nonenal. The mRNA for 3HBD was ubiquitously expressed in rabbit tissues. The results suggest that 3HBD is an NADPH-preferring reductase, and plays roles in the metabolisms of steroids, prostaglandin D₂, carbohydrates and xenobiotics, as well as a defense system, protecting against reactive carbonyl compounds.
Product Description
Supelco
Hexobarbital solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate hydrate
Sigma-Aldrich
Phenolphthalein solution, 0.5 wt. % in ethanol: water (1:1)
Sigma-Aldrich
Phenolphthalein, ACS reagent
Sigma-Aldrich
Isatin, 97%
Sigma-Aldrich
Prostaglandin D2, ≥95%, synthetic