Merck
  • In vitro effects of proteases in human pancreatic juice on stability of liquid and carrier-bound fibrin sealants.
In vitro effects of proteases in human pancreatic juice on stability of liquid and carrier-bound fibrin sealants.
The British journal of surgery (2013-09-17)
J Adelmeijer, R J Porte, T Lisman
ABSTRACT
Fibrin sealants are used in pancreatic surgery to prevent leakage of pancreatic fluid and reduce associated complications. The efficacy of this approach is unclear. Fibrin clots were generated in vitro from two commercially available liquid fibrin sealants (Tissucol Duo® and Evicel®) and the carrier-bound fibrin sealant Tachosil®, and exposed to normal saline or human pancreatic fluid. Stability of the sealants was assessed by release of the fibrin and collagen degradation products, D-dimer and hydroxyproline. The effect of protease inhibitors on sealant breakdown was assessed. Clots generated from liquid fibrin sealants degraded rapidly in pancreatic fluid, but not in normal saline. D-dimer release from fibrin clots by pancreatic fluid was approximately 1700 µg/ml after 24 h and less than 20 µg/ml by saline. Pancreatic fluid, but not normal saline, degraded both the fibrin and collagen component of Tachosil®. After 6 h, mean(s.e.m.) D-dimer levels in pancreatic fluid exposed to Tachosil® were 850(183) ng/ml, compared with 60(6) ng/ml in normal saline. The mean(s.e.m.) hydroxyproline concentration in pancreatic fluid was 497(17) µg/ml after a 24-h exposure to Tachosil®, compared with 26(12) µg/ml in normal saline. Protease inhibitors significantly inhibited breakdown of liquid sealants (D-dimer levels less than 50 µg/ml after 24 h) and Tachosil® (D-dimer release 179(12) ng/ml at 6 h; hydroxyproline release 181(29) µg/ml at 24 h). Proteases in pancreatic juice effectively degrade both liquid and carrier-bound fibrin sealants in vitro. The use of these products in pancreatic surgery with the aim of preventing leakage of pancreatic fluid is not supported by this experimental study.
Sigma-Aldrich
Fibrinogen from human plasma, 35-65% protein (≥90% of protein is clottable).
Sigma-Aldrich
Fibrin from human plasma, insoluble powder
Sigma-Aldrich
Fibrinogen from rat plasma, 60-80% protein (≥60% of protein is clottable)
Sigma-Aldrich
Fibrinogen from human plasma, 50-70% protein (≥80% of protein is clottable)
Sigma-Aldrich
trans-4-Hydroxy-L-proline, ≥99%
Sigma-Aldrich
trans-4-Hydroxy-L-proline, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
trans-4-Hydroxy-L-proline, BioReagent, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Fibrinogen from bovine plasma, Type I-S, 65-85% protein (≥75% of protein is clottable)
Supelco
trans-4-Hydroxy-L-proline, analytical standard