Merck
  • Nicotinamide, a glucose-6-phosphate dehydrogenase non-competitive mixed inhibitor, modifies redox balance and lipid accumulation in 3T3-L1 cells.
Nicotinamide, a glucose-6-phosphate dehydrogenase non-competitive mixed inhibitor, modifies redox balance and lipid accumulation in 3T3-L1 cells.
Life sciences (2013-11-05)
Nayeli Torres-Ramírez, Luis Arturo Baiza-Gutman, Rebeca García-Macedo, Clara Ortega-Camarillo, Alejandra Contreras-Ramos, Rafael Medina-Navarro, Miguel Cruz, Miguel Ángel Ibáñez-Hernández, Margarita Díaz-Flores
ABSTRACT
Excessive energy uptake of dietary carbohydrates results in their storage as fat and requires glucose-6-phosphate dehydrogenase (G6PD)-mediated NADPH production. We sought to assess whether the nicotinamide-induced reduction of G6PD activity might modulate redox balance and lipid accumulation in 3T3-L1 cells. 3T3-L1 preadipocytes (days 4 and 6 of differentiation) and adipocytes were cultured in the presence of 5 or 25 mM glucose. The cells cultured in 25 mM glucose were supplemented with nicotinamide (5-15 mM). Next, we evaluated the following parameters: cell viability, apoptosis, lipid accumulation, lipolysis, reducing power, reactive oxygen species (ROS), NAD(P)H and NAD(P)(+), isocitrate dehydrogenase (IDP), malic enzyme and G6PD, as well as the protein and mRNA levels of G6PD. We also analysed the kinetics of the nicotinamide-induced inhibition of G6PD. G6PD mRNA levels increased at day 4 of adipocyte differentiation, whereas G6PD activity progressively increased at days 4 and 6 of differentiation and was reduced in adipocytes. Concomitantly, ROS, reducing power and lipid accumulation increased gradually as the preadipocytes matured into adipocytes. High glucose increased the activity of G6PD, which coincided with an increase in ROS, reducing power and lipid accumulation. All of these changes are prevented by nicotinamide, with the exception of lipid accumulation in adipocytes. Nicotinamide increased IDP activity without affecting NADPH levels. Lastly, nicotinamide inhibited G6PD in a non-competitive mixed way. Nicotinamide modulates G6PD via a non-competitive mixed inhibition and decreases high glucose-dependent oxidative stress and lipid accumulation. Nicotinamide maintains NADPH levels by increasing the activity of IDP.
Product Description
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Nicotinamide (Niacinamide), analytical standard
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type IX, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type XV, lyophilized powder, 200-400 units/mg protein (modified Warburg-Christian)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from baker′s yeast (S. cerevisiae), Type VII, ammonium sulfate suspension, ≥200 units/mg protein
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Glycerol, FCC, FG
Supelco
Nicotinamide (Vitamin B3) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, Type XXIII, ammonium sulfate suspension, 550-1,100 units/mg protein (biuret), ≥2.0 mg/mL Biuret
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, recombinant, expressed in E. coli, lyophilized powder, ≥550 units/mg protein (biuret)
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, lyophilized powder, >= 550 units/mg protein (biuret)
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Glucose-6-phosphate Dehydrogenase from Leuconostoc mesenteroides, recombinant, expressed in E. coli, ammonium sulfate suspension, ≥550 units/mg protein (biuret)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide phosphate hydrate
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%