Merck
  • Highly dispersed ruthenium nanoparticle-embedded mesoporous silica as a catalyst for the production of gamma-butyrolactone from succinic anhydride.
Highly dispersed ruthenium nanoparticle-embedded mesoporous silica as a catalyst for the production of gamma-butyrolactone from succinic anhydride.
Journal of nanoscience and nanotechnology (2013-11-20)
Sang-Ho Chung, Hee-Jun Eom, Min-Sung Kim, Myung Suk Lee, Kwan-Young Lee
ABSTRACT
In this study, a novel, strategic method was developed for the synthesis of a mesoporous silica catalyst embedded with ruthenium nanoparticles (RuNPs/SiO2) by combining the polyol and modified sol-gel methods. By applying this new procedure, uniformly synthesized ruthenium nanoparticles with an average size of 3.8 nm and 95% spherical shape were highly dispersed in the mesoporous silica support material. Coordinated carbonyl groups of PVP remaining from the synthesis of the RuNPs were effectively removed by the thermal treatment (calcined at 573 K for 4 h) and the sythesized RuNPs/SiO2 catalysts were reduced under hydrogen at 20 bar for 2 h. These catalysts were analyzed using transmission electron microscopy (TEM), Fourier transform infrared spectroscopy (FT-IR), N2 adsorption-desorption, and X-ray diffraction (XRD). After the thermal treatment and the reduction procedure, the size and shape of the embedded RuNPs were nearly unchanged, and the catalyst was active in the liquid-phase hydrogenation of succinic anhydride (SAN) to selectively form y-butyrolactone (GBL) with a maximum yield of 90.1%. This novel catalyst preparation is a potentially useful method for the synthesis of metal nanoparticles as heterogeneous catalysts.
Sigma-Aldrich
Succinic anhydride, ≥99% (GC)
Sigma-Aldrich
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O
Sigma-Aldrich
γ-Butyrolactone, ReagentPlus®, ≥99%
Sigma-Aldrich
Ruthenium, powder, 99.99% trace metals basis
Sigma-Aldrich
Ruthenium, powder, −200 mesh, 99.9% trace metals basis
Sigma-Aldrich
4-Hydroxybutanoic acid lactone, ≥98%, FCC, FG
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
Silica
Sigma-Aldrich
Silicon dioxide, alumina doped, nanoparticles, dispersion, <50 nm particle size, 20 wt. % in H2O, ≥99.9% trace metals basis
Sigma-Aldrich
Silica, mesostructured, MSU-F (cellular foam)
Sigma-Aldrich
Silica, mesostructured, MCM-41 type (hexagonal)
Sigma-Aldrich
Silica, nanopowder, 99.8% trace metals basis
Sigma-Aldrich
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Sigma-Aldrich
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Sigma-Aldrich
Ruthenium black
Sigma-Aldrich
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis