Merck
  • Potential benefit of telbivudine on renal function does not outweigh its high rate of antiviral drug resistance and other adverse effects.
Potential benefit of telbivudine on renal function does not outweigh its high rate of antiviral drug resistance and other adverse effects.
Gastroenterology (2013-11-28)
Suna Yapali, Anna S Lok
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Thymidine, ≥99%