Skip to Content
PL
EN
Products
Applications
Services
Resources
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Lab Automation
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Documents
Learning Center
New
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Home
Search Results
ECG response: November 26, 2013.
ECG response: November 26, 2013.
Circulation (2013-11-28)
PMID
24276877
Product Description
A9251
Sigma-Aldrich
Adenosine, ≥99%
Pricing
A4036
Sigma-Aldrich
Adenosine, BioReagent, suitable for cell culture
Pricing
A0230200
Adenosine, European Pharmacopoeia (EP) Reference Standard
Pricing
PHR1138
Supelco
Adenosine, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing
01890
Sigma-Aldrich
Adenosine
Pricing