- [Myeloperoxidase in the neurodegenerative process of Parkinson's disease].
[Myeloperoxidase in the neurodegenerative process of Parkinson's disease].
Deutsche medizinische Wochenschrift (1946) (2013-11-28)
P Teismann
PMID24277448
ABSTRACT
Myeloperoxidase (MPO) is a hemoprotein which is involved in the unspecific immune response. In this process hypochloric acid is released. Hypochloric acid can react with lipids and proteins and thus lead to cell damage. We were able to show that MPO, as well as a biomarker for MPO - 3-Chlorotyrosine - are upregulated in the 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP)-model of Parkinson's disease as well as in the disease itself. Ablation of MPO resulted in a neuroprotective effect in the MPTP-model. Herein we describe the different function of MPO, and how these can lead to the cellular demise as seen in Parkinson's disease.
Product Description
Dopamine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Dopamine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dopamine hydrochloride solution, 1.0 mg/mL in methanol with 5% 1 M HCl (as free base), ampule of 1 mL, certified reference material, Cerilliant®