Merck
  • Pharmacokinetic and pharmacodynamic properties of enrofloxacin in southern crested caracaras (Caracara plancus).
Pharmacokinetic and pharmacodynamic properties of enrofloxacin in southern crested caracaras (Caracara plancus).
Journal of avian medicine and surgery (2013-12-19)
Samanta Waxman, Ana Paula Prados, Julio de Lucas, Manuel Ignacio San Andres, Juan Carlos Sassaroli, Marcela Orozco, Hernán Argibay, Casilda Rodriguez
ABSTRACT
To determine the dosage of enrofloxacin in southern crested caracaras (Caracara plancus), plasma concentrations of enrofloxacin were measured by high-performance liquid chromatography after intravenous (IV) (5 mg/kg) and intramuscular (IM) (10 mg/kg) administration. This compound presented a relatively high volume of distribution (2.09 L/kg), a total body clearance of 0.24 L/kg x h, and a long permanence as shown by an elimination half-life of 7.81 hours after IV administration and a terminal half-life of 6.58 hours after IM administration. The areas under the concentration-time curves (AUC) were 21.92 and 34.38 microg x h/mL for IM and IV administration, respectively. Enrofloxacin was rapidly absorbed after IM administration with a time to reach maximum concentration of 0.72 hours and bioavailability of 78.76%. After IM administration, the peak drug concentration (C(max)) was 3.92 microg/mL. Values of minimum inhibitory concentration (MIC), C(max), and AUC have been used to predict the clinical efficacy of a drug in treating bacterial infections, with a C(max)/MIC value of 10 and an AUC/MIC ratio of 125-250 associated with optimal bactericidal effects. By using the study data and a MIC breakpoint of 0.25 microg/mL, values of C(max)/MIC were 13.74 and 15.94 and for AUC/MIC were 90.73 and 139.63, for the IV and IM routes respectively. For the treatment of infectious diseases caused by microorganisms with MIC < or = 0.25 microg/mL, the calculated optimal dosages were 7.5 and 9.5 mg/kg q24h by the IV and IM routes, respectively. For less susceptible bacteria, a dose increase should be evaluated. To treat caracara by the IV route against microorganisms with MIC < or = 0.25 microg/mL, the dose should be higher than the 5 mg/kg used in our study, but possible side effects derived from an increase in the IV dose and efficacy in sick birds should be assessed.
Product Description
Supelco
Ciprofloxacin HCl, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Supelco
Enrofloxacin, VETRANAL®, analytical standard
Supelco
Ciprofloxacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Enrofloxacin, ≥99.0%
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Enrofloxacin, European Pharmacopoeia (EP) Reference Standard
Ciprofloxacin hydrochloride for peak identification, European Pharmacopoeia (EP) Reference Standard
Ciprofloxacin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Ciprofloxacin, European Pharmacopoeia (EP) Reference Standard