Merck
  • Ascorbic acid reverses the prolonged anesthetic action of pentobarbital in Akr1a-knockout mice.
Ascorbic acid reverses the prolonged anesthetic action of pentobarbital in Akr1a-knockout mice.
Life sciences (2013-12-21)
Junitsu Ito, Noriyuki Otsuki, Xuhong Zhang, Tasuku Konno, Toshihiro Kurahashi, Motoko Takahashi, Mayumi Yamato, Yuta Matsuoka, Ken-ichi Yamada, Satoshi Miyata, Junichi Fujii
ABSTRACT
Aldehyde reductase (AKR1A), a member of the aldo-keto reductase superfamily, is highly expressed in the liver and is involved in both the detoxification of carbonyl compounds and ascorbic acid biosynthesis. By comparison with wild-type mice, Akr1a-knockout (Akr1a(-/-)) mice and human Akrla-transgenic (Akr1a(tg/+)) mice experience different anesthetic actions from pentobarbital-prolonged in Akr1a-knockout (Akr1a(-/-)) mice and shortened in human Akrla-transgenic (Akr1a(tg/+)) mice. We investigated this alteration in the anesthetic efficacy of pentobarbital in Akr1a genetically modified mice. Neither the cytosolic protein of wild-type mouse liver nor purified rat AKR1A directly reduced pentobarbital. Ascorbic acid administration neutralized the prolonged duration of the loss of the righting reflex (LORR) in Akr1a(-/-) mice, but preincubation of pentobarbital with ascorbic acid prior to administration did not change the anesthetic effect. Those results indicated that ascorbic acid does not directly reduce pentobarbital. Enzymatic activities and levels of the proteins of some cytochrome P450s that make up a potent detoxification system for pentobarbital showed no changes in the genetically modified mice examined. Thus, ascorbic acid also had no effect on the detoxification system in the liver. The prolonged duration of LORR in the Akr1a(-/-) mice caused by pentobarbital and the neutralization of the anesthetic effect by ascorbic acid together with other results imply that ascorbic acid alters the responses of the neuronal system to anesthetics. Pentobarbital action is increased under conditions of ascorbic acid deficiency, and this may have to be taken into account when anesthetizing malnourished patients.
Product Description
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Sodium ascorbate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Supelco
Pentobarbital solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%