Merck
Measuring membrane protein stability under native conditions.
Proceedings of the National Academy of Sciences of the United States of America (2013-12-25)
Yu-Chu Chang, James U Bowie
ABSTRACT
The thermodynamic stability of proteins is typically measured at high denaturant concentrations and then extrapolated back to zero denaturant conditions to obtain unfolding free energies under native conditions. For membrane proteins, the extrapolations are fraught with considerable uncertainty as the denaturants may have complex effects on the membrane or micellar structure. We therefore sought to measure stability under native conditions, using a method that does not perturb the properties of the membrane or membrane mimetics. We use a technique called steric trapping to measure the thermodynamic stability of bacteriorhodopsin in bicelles and micelles. We find that bacteriorhodopsin has a high thermodynamic stability, with an unfolding free energy of ∼11 kcal/mol in dimyristoyl phosphatidylcholine bicelles. Nevertheless, the stability is much lower than predicted by extrapolation of measurements made at high denaturant concentrations. We investigated the discrepancy and found that unfolding free energy is not linear with denaturant concentration. Apparently, long extrapolations of helical membrane protein unfolding free energies must be treated with caution. Steric trapping, however, provides a method for making these measurements.
Product Description
Supelco
Biotin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Biotin, tested according to Ph. Eur.
Supelco
Biotin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Biotin, meets USP testing specifications
Biotin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Biotin, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Biotin, ≥99.0% (T)