Merck
  • The use of calcium phosphate cement in vertebroplasty of the base of odontoid process.
The use of calcium phosphate cement in vertebroplasty of the base of odontoid process.
Neurologia i neurochirurgia polska (2014-01-01)
Krzysztof Zapałowicz, Maciej Wojdyn, Krzysztof Włodzimierz Zieliński, Dorota Snopkowska-Wiaderna
ABSTRACT
The authors describe the use of bone cement containing calcium phosphate for vertebroplasty of the cavity in the base of odontoid process. A 23-year-old female patient was operated on by incision in lateral cervical area (anterior open access). After a blunt dissection, the working cannula (Kyphon) was introduced under fluoroscopic guidance through the C2 vertebral body to the cavity in the base of the odontoid process. Intraoperatively, biopsy of the lesion was taken and histo-pathological examination excluded the presence of neoplasm. The cavity, presumably haemangioma, was successfully filled with calcium phosphate bone cement KyphOsTM FS (Ky-phon). The proper filling without paravertebral cement leak was confirmed by postoperative computed tomography (CT). The CT and magnetic resonance imaging performed 9 months after the procedure showed that cement was still present in the cavity. This is the first use of calcium phosphate cement to conduct the vertebroplasty of C2 vertebra.
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), contains 5 wt. % silica as dopant, synthetic
Sigma-Aldrich
Hydroxyapatite, nanoparticles, dispersion, 10 wt. % in H2O, <200 nm particle size (BET)
Sigma-Aldrich
Hydroxyapatite, synthetic, 99.8% trace metals basis (excludes Mg)
Sigma-Aldrich
Calcium phosphate tribasic, 34.0-40.0% Ca basis
Sigma-Aldrich
Calcium phosphate tribasic, BioReagent, suitable for plant cell culture, powder
Sigma-Aldrich
Hydroxyapatite, nanopowder, <200 nm particle size (BET), ≥97%, synthetic
Sigma-Aldrich
Hydroxyapatite, powder, synthetic