Merck
  • The relation between cryoprotective and physicochemical properties of oxyethylated methyl cellosolve-based media.
The relation between cryoprotective and physicochemical properties of oxyethylated methyl cellosolve-based media.
Cryo letters (2014-01-23)
A V Nikolenko, V V Chekanova, M I Schetinsky, O V Vyazovska
ABSTRACT
Oxyethyl derivatives of polyol class members such as glycerol and ethylene glycol with various polymerization degrees possess low toxicity and cryoprotective properties via an exocellular mechanism. To develop the cryoprotective media based on non-penetrating cryoprotectants, we studied cryoprotective and physico-chemical properties of oxyethylated methyl cellosolve (OEMC)-based media. Media containing 20% and 30% OEMC were prepared with 50 mmol/l and 150 mmol/l NaCl. In some media, NaCl content was reduced, and sucrose, mannitol or glucose was added to attain the isotonic media. Cryopreservation of red blood cells depends on the media composition and physico-chemical properties. The increases in dynamic viscosity due to higher OEMC concentration and in ionic strength resulted from higher NaCl concentration lead to greater erythrocytes dehydration prior to freezing, and lower red blood cell preservation upon freezing. Salt substitution with carbohydrates in 20% OEMC media reduces ionic strength and increases OEMC distribution coefficient. The correlation of red blood cell protection upon freezing is discussed in terms of the media physico-chemical properties (ionic strength, viscosity and surface tension), cryoprotectant distribution coefficient, and cell dehydration.
Product Description
Supelco
Ethylene glycol solution, suitable for NMR (reference standard), 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Supelco
Ethylene glycol, analytical standard
Supelco
2-Methoxyethanol, analytical standard
Sigma-Aldrich
2-Methoxyethanol, anhydrous, 99.8%
Sigma-Aldrich
Ethylene glycol, ReagentPlus®, ≥99%
Supelco
Ethylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Methoxyethanol, contains 50 ppm BHT as stabilizer, ACS reagent, ≥99.3%
Sigma-Aldrich
Ethylene glycol, spectrophotometric grade, ≥99%
Sigma-Aldrich
2-Methoxyethanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
2-Methoxyethanol, ReagentPlus®, ≥99.0%, contains 50 ppm BHT as stabilizer
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylene glycol 5 M solution
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%