Merck
  • Confluent hepatic fibrosis as the presenting imaging sign in nonadvanced alcoholic cirrhosis.
Confluent hepatic fibrosis as the presenting imaging sign in nonadvanced alcoholic cirrhosis.
Clinical imaging (2004-03-31)
Nikolaos L Kelekis, Eirini Makri, Aikaterini Vassiou, Kalliopi Patsiaoura, Michalis Spiridakis, Georgios N Dalekos
ABSTRACT
Confluent hepatic fibrosis (CHF) is usually a feature of advanced cirrhosis, while occurrence in early stage compensated cirrhosis is uncommon. We report the imaging findings of masslike CHF in a patient with previously unsuspected compensated alcoholic cirrhosis. The combination of partially wedge shape, minimal capsular retraction, and homogeneous delayed enhancement on multiphase contrast-enhanced spiral CT led to the characterization of the lesions and establishment of the diagnosis of cirrhosis.
Product Description
USP
Iodixanol Related Compound D, United States Pharmacopeia (USP) Reference Standard
Iopentol, European Pharmacopoeia (EP) Reference Standard