Merck
  • [Application of nootropic agents in complex treatment of patients with concussion of the brain].
[Application of nootropic agents in complex treatment of patients with concussion of the brain].
Likars'ka sprava (2008-04-18)
A V Tkachev
ABSTRACT
65 patients with a mild craniocereberal trauma have been observed. Medical examination included among general clinical methods the following methods: KT (MRT) of the brain, oculist examination including the observation of eye fundus. For objectification of a patient' complaints the authors used orientation and Galvestona's amnesia tests, feeling scale (psychological test), the table to determine the level of memory. Tests have been carried out on the first, tenth and thirty day of the treatment. Patients of the first group received in a complex treatment -pramistar, patients of the second group - piracetam. Patients of both groups noted considerable improvement during a complex treatment (disappearance of headache, dizziness and nausea) and at the same time patients receiving pramistar had better restoration of orientation and feeling. Pramistar was also more effective in patients with amnesia.
Sigma-Aldrich
Pramiracetam, ≥98% (HPLC)