Merck
  • Associations of single nucleotide polymorphisms with malignant and borderline bone tumors.
Associations of single nucleotide polymorphisms with malignant and borderline bone tumors.
Klinicheskaia laboratornaia diagnostika (2014-03-20)
N E Kushlinskii, Yu S Tivofeev, E V Generozov, V A Naumov, Yu N Soloviev, I V Boulytcheva, M D Aliev
ABSTRACT
Bone neoplasms - are a rare group of diseases, which ethiology and pathogenesis are not fully understood. We have studied 6 single nucleotide polymorphisms rs792/(GHI), rs7956547(IGFI), rs3761243(GNRH2), rs11737764(FGF2), rs6599400(FGFR3), and rs1690916(MDM2) associations with bone tumors. In our work we've detected significant associations with some single nucleotide polymorphisms: IGFl.rs7956547, GNRH2.rs3761243 and FGFR3.rs6599400 in patients with malignant and borderline bone tumors.
Product Description
Gonadorelin, European Pharmacopoeia (EP) Reference Standard