Merck
  • Pharmacologic therapeutic window of pramiracetam demonstrated in behavior, EEG, and single neuron firing rates.
Pharmacologic therapeutic window of pramiracetam demonstrated in behavior, EEG, and single neuron firing rates.
Experientia (1985-09-15)
B P Poschel, P M Ho, F W Ninteman, M J Callahan
ABSTRACT
Following oral or intravenous administration, a representative cognition activator drug, pramiracetam sulfate, is shown to have a pharmacologic therapeutic window at three different levels of study: learned behavior, gross EEG activity of the frontal cortex and hippocampus, and firing rate of single hippocampal neurons.
Sigma-Aldrich
Pramiracetam, ≥98% (HPLC)