Merck
  • Patients with Fabry disease after enzyme replacement therapy dose reduction versus treatment switch.
Patients with Fabry disease after enzyme replacement therapy dose reduction versus treatment switch.
Journal of the American Society of Nephrology : JASN (2014-02-22)
Frank Weidemann, Johannes Krämer, Thomas Duning, Malte Lenders, Sima Canaan-Kühl, Alice Krebs, Hans Guerrero González, Claudia Sommer, Nurcan Üçeyler, Markus Niemann, Stefan Störk, Michael Schelleckes, Stefanie Reiermann, Jörg Stypmann, Stefan-Martin Brand, Christoph Wanner, Eva Brand
ABSTRACT
Because of the shortage of agalsidase-beta in 2009, many patients with Fabry disease were treated with lower doses or were switched to agalsidase-alfa. This observational study assessed end-organ damage and clinical symptoms during dose reduction or switch to agalsidase-alfa. A total of 105 adult patients with Fabry disease who had received agalsidase-beta (1.0 mg/kg body weight) for ≥1 year were nonrandomly assigned to continue this treatment regimen (regular-dose group, n=38), receive a reduced dose of 0.3-0.5 mg/kg (dose-reduction group, n=29), or switch to 0.2 mg/kg agalsidase-alfa (switch group) and were followed prospectively for 1 year. We assessed clinical events (death, myocardial infarction, severe arrhythmia, stroke, progression to ESRD); changes in cardiac, renal, and neurologic function; and Fabry-related symptoms (neuropathic pain, hypohidrosis, diarrhea, and disease severity scores). Organ function and Fabry-related symptoms remained stable in the regular-dose group. In contrast, estimated GFR decreased by about 3 ml/min per 1.73 m(2) (P=0.01) in the dose-reduction group, and the median albumin-to-creatinine ratio increased from 114 (0-606) mg/g to 216 (0-2062) mg/g (P=0.03) in the switch group. Furthermore, mean Mainz Severity Score Index scores and frequencies of pain attacks, chronic pain, gastrointestinal pain, and diarrhea increased significantly in the dose-reduction and switch groups. In conclusion, patients receiving regular agalsidase-beta dose had a stable disease course, but dose reduction led to worsening of renal function and symptoms. Switching to agalsidase-alfa is safe, but microalbuminuria may progress and Fabry-related symptoms may deteriorate.
Sigma-Aldrich
α-Galactosidase from green coffee beans, ammonium sulfate suspension, ≥9 units/mg protein
Sigma-Aldrich
α-Galactosidase, positionally specific from Escherichia coli, recombinant, expressed in E. coli, buffered aqueous solution