Merck
  • A sensitive and robust method for automated on-line monitoring of enzymatic activities in water and water resources.
A sensitive and robust method for automated on-line monitoring of enzymatic activities in water and water resources.
Water science and technology : a journal of the International Association on Water Pollution Research (2014-03-22)
G Ryzinska-Paier, T Lendenfeld, K Correa, P Stadler, A P Blaschke, R L Mach, H Stadler, A K T Kirschner, A H Farnleitner
ABSTRACT
The realisation of a novel concept for automated on-line monitoring of enzymatic activities in water was successfully demonstrated by long-term field testing at two remote Austrian ground water resources. The β-D-glucuronidase (GLUC) activity was selected as a representative enzymatic model parameter for the on-line determination. But the device can be adapted for any enzymatic reaction with diagnostic relevance for microbial water quality monitoring, as demonstrated for the β-D-galactosidase activity. Automated filtration of volumes up to 5 litres supports sensitive quantification of enzymatic activities. Internet-based data transfer, using internal control parameters for verification and a dynamic determination of the limit of quantification, enabled robust enzymatic on-line monitoring during a 2-year period. A proportion of 5,313 out of 5,506 GLUC activity measurements (96.5%) could be positively verified. Hydrological (discharge, gauge, turbidity, temperature, pH, electric conductivity, spectral absorbance coefficient at 254 nm) as well as microbiological parameters (Escherichia coli, coliforms) were concurrently determined to characterise the investigated ground water resources. The enzymatic on-line measurements closely reflected the different hydrological conditions and contamination patterns of the test sites. Contrary to expectations, GLUC did not qualify as a proxy-parameter for the occurrence of cultivation-based E. coli contamination and warrants further detailed investigations on its indication capacity as a rapid means for microbial faecal pollution detection in such aquatic habitats. Microbial on-line monitoring is likely to become more important in the future, complementing existing surveillance strategies for water safety management. Further perspectives on the application of such analytical on-line technologies, such as their connection with event-triggered sampling and standardised diagnostics, are discussed.
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-2, aqueous solution, ≥85,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type HP-2, aqueous solution, ≥100,000 units/mL
Sigma-Aldrich
β-Glucuronidase from limpets (Patella vulgata), Type L-II, lyophilized powder, 1,000,000-3,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-3AF, aqueous solution, ≥60,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-3, aqueous solution, ≥90,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type HP-2S, aqueous solution, ≥90,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-5, lyophilized powder, ≥400,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-1, partially purified powder, ≥300,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from limpets (Patella vulgata), aqueous solution, ≥85,000 units/mL
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, Type IX-A, lyophilized powder, 1,000,000-5,000,000 units/g protein (30 min assay)
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, ≥10,000,000 units/g protein (30 min assay), recombinant, expressed in E. coli overproducing strain, lyophilized powder
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, ≥20,000 units/mg protein, recombinant, expressed in E. coli overproducing strain, lyophilized powder
Sigma-Aldrich
β-Glucuronidase from bovine liver, Type B-3, ≥2,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, aqueous glycerol solution, ≥5,000,000 units/g protein, pH 6.8 (biuret)
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, Type VII-A, lyophilized powder, 5,000,000-20,000,000 units/g protein, pH 6.8 (30 min assay)
Sigma-Aldrich
β-Glucuronidase from bovine liver, Type B-1, ≥1,000,000 units/g solid
Sigma-Aldrich
β-Glucuronidase from Helix aspersa (garden snail), Type HA-4
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, >20,000,000 units/g protein, recombinant, expressed in E. coli, aqueous glycerol solution